Assay Details
| Gene Symbol: SC5D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689562.3 | XP_005689619.2 | 3 - 4 | 480 | 92 | |
| GenBank mRNA | JO634883.2 | - | 3 - 4 | 501 | 92 | |
More Information
Set Membership: |
|

SC5D
Genomic Map