Assay Details
| Gene Symbol: C17H22orf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691739.3 | XP_005691796.2 | 5 - 6 | 675 | 77 | |
| XM_018060913.1 | XP_017916402.1 | 5 - 6 | 784 | 77 | ||
More Information
Set Membership: |
|

C17H22orf15
Genomic Map