Assay Details
Gene Symbol: CMTM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692106.3 | XP_005692163.1 | 1 - 2 | 395 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
