Assay Details
| Gene Symbol: TXNL4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039392.1 | XP_017894881.1 | 3 - 4 | 402 | 72 | |
| XM_018039393.1 | XP_017894882.1 | 3 - 4 | 384 | 72 | ||
| XM_018039394.1 | XP_017894883.1 | 4 - 5 | 458 | 72 | ||
| XM_018039395.1 | XP_017894884.1 | 4 - 5 | 354 | 72 | ||
| GenBank mRNA | JO446908.2 | - | 3 - 4 | 386 | 72 | |
| JO644141.2 | - | 3 - 4 | 417 | 72 | ||
More Information
Set Membership: |
|

TXNL4A
Genomic Map