Assay Details
| Gene Symbol: GGPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042543.1 | XP_017898032.1 | 3 - 4 | 385 | 84 | |
| GenBank mRNA | JO447546.2 | - | 1 - 2 | 400 | 84 | |
More Information
Set Membership: |
|

GGPS1
Genomic Map