Assay Details
Gene Symbol: FRRS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018052565.1 | XP_017908054.1 | 3 - 4 | 610 | 60 | |
XM_018052566.1 | XP_017908055.1 | 3 - 4 | 492 | 60 | ||
XM_018052567.1 | XP_017908056.1 | 3 - 4 | 492 | 60 | ||
XM_018052568.1 | XP_017908057.1 | 3 - 4 | 492 | 60 | ||
GenBank mRNA | JT649783.1 | - | 3 - 4 | 544 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
