Assay Details
| Gene Symbol: LOC102182436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013971736.2 | XP_013827190.2 | 3 - 4 | 793 | 72 | |
| XM_013971738.2 | XP_013827192.2 | 2 - 3 | 218 | 72 | ||
| XM_013971739.1 | XP_013827193.1 | 1 - 2 | 1133 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013971738.2 transcripts.
Set Membership: |
|

LOC102182436
Genomic Map