Assay Details
| Gene Symbol: LOC102177672 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013972752.2 | XP_013828206.2 | 5 - 6 | 1334 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102177672| Gene Symbol: LOC102177672 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013972752.2 | XP_013828206.2 | 5 - 6 | 1334 | 92 | |
Set Membership: |
|