Assay Details
| Gene Symbol: NFE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679848.3 | XP_005679905.2 | 2 - 3 | 364 | 68 | |
| GenBank mRNA | JO340606.2 | - | 2 - 3 | 433 | 68 | |
More Information
Set Membership: |
|

NFE2
Genomic Map