Assay Details
Gene Symbol: PPBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681750.3 | XP_005681807.2 | 2 - 3 | 299 | 62 | |
GenBank mRNA | JO330674.2 | - | 2 - 3 | 370 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
