Assay Details
| Gene Symbol: ADRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683143.3 | XP_005683200.1 | 1 | 1434 | 88 | |
| GenBank mRNA | JT620058.1 | - | 1 | 1223 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: ADRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683143.3 | XP_005683200.1 | 1 | 1434 | 88 | |
| GenBank mRNA | JT620058.1 | - | 1 | 1223 | 88 | |
Set Membership: |
|