Assay Details
Gene Symbol: SHARPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058712.1 | XP_017914201.1 | 4 - 5 | 1094 | 69 | |
GenBank mRNA | JO630195.2 | - | 4 - 5 | 1087 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
