Assay Details
| Gene Symbol: GRINA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058698.1 | XP_017914187.1 | 4 - 5 | 868 | 59 | |
| GenBank mRNA | JO363369.2 | - | 4 - 5 | 866 | 59 | |
| JO431649.2 | - | 4 - 5 | 853 | 59 | ||
More Information
Set Membership: |
|

GRINA
Genomic Map