Assay Details
Gene Symbol: PAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689126.3 | XP_005689183.2 | 6 - 7 | 608 | 57 | |
XM_013969305.2 | XP_013824759.2 | 5 - 6 | 531 | 57 | ||
GenBank mRNA | JO329930.2 | - | 6 - 7 | 594 | 57 | |
JO585835.2 | - | 6 - 7 | 797 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
