Assay Details
| Gene Symbol: MIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692421.3 | XP_005692478.1 | 2 - 3 | 2655 | 59 | |
| XM_005692423.3 | XP_005692480.1 | 3 - 4 | 1332 | 59 | ||
More Information
Set Membership: |
|

MIA
Genomic Map