Assay Details
Gene Symbol: METTL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005694106.3 | XP_005694163.1 | 4 - 5 | 816 | 86 | |
XM_013972345.2 | XP_013827799.1 | 3 - 4 | 400 | 86 | ||
GenBank mRNA | JO595594.2 | - | 4 - 5 | 789 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
