Assay Details
Gene Symbol: C23H6orf89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696265.3 | XP_005696322.2 | 3 - 4 | 426 | 68 | |
XM_005696266.3 | XP_005696323.2 | 2 - 3 | 353 | 68 | ||
XM_013973851.2 | XP_013829305.2 | 3 - 4 | 654 | 68 | ||
GenBank mRNA | JO424772.1 | - | 3 - 4 | 464 | 68 | |
KA336915.1 | - | 6 - 7 | 4824 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
