Assay Details
Gene Symbol: C23H6orf89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696265.3 | XP_005696322.2 | 8 - 9 | 1194 | 113 | |
XM_005696266.3 | XP_005696323.2 | 7 - 8 | 1121 | 113 | ||
XM_013973851.2 | XP_013829305.2 | 8 - 9 | 1422 | 113 | ||
XM_013973852.2 | XP_013829306.2 | 6 - 7 | 814 | 113 | ||
GenBank mRNA | JO424772.1 | - | 8 - 9 | 1232 | 113 | |
KA336915.1 | - | 1 - 2 | 4056 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
