Assay Details
| Gene Symbol: TXNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039606.1 | XP_017895095.1 | 4 - 5 | 754 | 69 | |
| XM_018039607.1 | XP_017895096.1 | 4 - 5 | 754 | 69 | ||
| XM_018039608.1 | XP_017895097.1 | 4 - 5 | 754 | 69 | ||
More Information
Set Membership: |
|

TXNL1
Genomic Map