Assay Details
Gene Symbol: LOC108634897 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045228.1 | XP_017900717.1 | 562 | 104 | ||
GenBank mRNA | JO436090.2 | - | 562 | 104 |
Gene Symbol: FAM120C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043858.1 | XP_017899347.1 | 12 - 13 | 2730 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
