Assay Details
Gene Symbol: LOC108633216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045500.1 | XP_017900989.1 | 5 - 6 | 729 | 86 |
Gene Symbol: LOC108634865 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045210.1 | XP_017900699.1 | 5 - 6 | 495 | 86 |
Gene Symbol: LOC108635049 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045352.1 | XP_017900841.1 | 3 - 4 | 438 | 86 |
Gene Symbol: LOC108635343 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045525.1 | XP_017901014.1 | 9 - 10 | 1413 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
