Assay Details
| Gene Symbol: LOC108635343 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045525.1 | XP_017901014.1 | 67 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018045525.1 transcripts.
Set Membership: |
|

LOC108635343
Genomic Map