Assay Details
| Gene Symbol: LOC102178217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066571.1 | XP_017922060.1 | 13 - 14 | 1375 | 93 | |
| GenBank mRNA | JO646020.2 | - | 13 - 14 | 1446 | 93 | |
| JO646021.2 | - | 13 - 14 | 1446 | 93 | ||
More Information
Set Membership: |
|

LOC102178217
Genomic Map