Assay Details
Gene Symbol: BLOC1S5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013974166.2 | XP_013829620.2 | 3 - 4 | 393 | 66 | |
GenBank mRNA | KA313478.1 | - | 3 - 4 | 292 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
