Assay Details
Gene Symbol: MAPK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039722.1 | XP_017895211.1 | 2 - 3 | 1552 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
