Assay Details
| Gene Symbol: RABGEF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040617.1 | XP_017896106.1 | 3 - 4 | 432 | 70 | |
| XM_018040618.1 | XP_017896107.1 | 3 - 4 | 429 | 70 | ||
| XM_018040619.1 | XP_017896108.1 | 2 - 3 | 260 | 70 | ||
| XM_018040620.1 | XP_017896109.1 | 2 - 3 | 294 | 70 | ||
| XM_018040621.1 | XP_017896110.1 | 2 - 3 | 294 | 70 | ||
| XM_018040622.1 | XP_017896111.1 | 1 - 2 | 187 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018040622.1 transcripts.
Set Membership: |
|

RABGEF1
Genomic Map