Assay Details
Gene Symbol: PRRG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044325.1 | XP_017899814.1 | 3 - 4 | 268 | 61 | |
XM_018044326.1 | XP_017899815.1 | 4 - 5 | 482 | 61 | ||
XM_018044327.1 | XP_017899816.1 | 3 - 4 | 405 | 61 | ||
GenBank mRNA | JO641570.2 | - | 1 - 2 | 620 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
