Assay Details
| Gene Symbol: CDKN2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678441.3 | XP_005678498.1 | 2 - 3 | 1337 | 90 | |
| XM_018044031.1 | XP_017899520.1 | 1 - 2 | 527 | 90 | ||
More Information
Set Membership: |
|

CDKN2C
Genomic Map