Assay Details
Gene Symbol: RHBDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678671.3 | XP_005678728.2 | 5 - 6 | 846 | 76 | |
XM_018042042.1 | XP_017897531.1 | 6 - 7 | 915 | 76 | ||
XM_018042057.1 | XP_017897546.1 | 6 - 7 | 915 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
