Assay Details
| Gene Symbol: MRPL42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679817.3 | XP_005679874.1 | 6 - 7 | 626 | 82 | |
| XM_005679818.3 | XP_005679875.1 | 6 - 7 | 621 | 82 | ||
| XM_018047624.1 | XP_017903113.1 | 6 - 7 | 654 | 82 | ||
| GenBank mRNA | JO515757.2 | - | 1 - 2 | 127 | 82 | |
| JO598400.2 | - | 1 - 2 | 127 | 82 | ||
More Information
Set Membership: |
|

MRPL42
Genomic Map