Assay Details
| Gene Symbol: CENPW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684480.2 | XP_005684537.1 | 2 - 3 | 441 | 98 | |
| GenBank mRNA | JO584837.2 | - | 1 - 2 | 294 | 98 | |
More Information
Set Membership: |
|

CENPW
Genomic Map