Assay Details
| Gene Symbol: LOC102175216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054405.1 | XP_017909894.1 | 13 - 14 | 1562 | 57 | |
| XM_018054406.1 | XP_017909895.1 | 13 - 14 | 1556 | 57 | ||
| XM_018054407.1 | XP_017909896.1 | 12 - 13 | 2043 | 57 | ||
| XM_018054408.1 | XP_017909897.1 | 14 - 15 | 1448 | 57 | ||
| XM_018054409.1 | XP_017909898.1 | 13 - 14 | 1929 | 57 | ||
| XM_018054411.1 | XP_017909900.1 | 18 - 19 | 1969 | 57 | ||
More Information
Set Membership: |
|

LOC102175216
Genomic Map