Assay Details
| Gene Symbol: LOC102175216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054410.1 | XP_017909899.1 | 3 - 4 | 510 | 72 | |
| XM_018054411.1 | XP_017909900.1 | 3 - 4 | 504 | 72 | ||
More Information
Set Membership: |
|

LOC102175216
Genomic Map