Assay Details
| Gene Symbol: LRRC8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055922.1 | XP_017911411.1 | 3 - 4 | 2402 | 88 | |
| XM_018055923.1 | XP_017911412.1 | 2 - 3 | 2266 | 88 | ||
| XM_018055924.1 | XP_017911413.1 | 4 - 5 | 2532 | 88 | ||
| GenBank mRNA | JO367595.2 | - | 739 | 88 | ||
| JO559106.2 | - | 2 - 3 | 2271 | 88 | ||
More Information
Set Membership: |
|

LRRC8A
Genomic Map