Assay Details
| Gene Symbol: RIMS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057912.1 | XP_017913401.1 | 5 - 6 | 655 | 58 | |
| GenBank mRNA | JO630683.2 | - | 1 - 2 | 1153 | 58 | |
More Information
Set Membership: |
|

RIMS4
Genomic Map