Assay Details
| Gene Symbol: HPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689766.3 | XP_005689823.1 | 9 - 10 | 1154 | 66 | |
| GenBank mRNA | JO421825.2 | - | 2 - 3 | 225 | 66 | |
More Information
Set Membership: |
|

HPX
Genomic Map