Assay Details
Gene Symbol: ATP6V0D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692458.3 | XP_005692515.2 | 6 - 7 | 1023 | 55 | |
GenBank mRNA | JO499679.2 | - | 2 - 3 | 666 | 55 | |
JO581431.1 | - | 6 - 7 | 984 | 55 | ||
JO581432.1 | - | 6 - 7 | 984 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
