Assay Details
Gene Symbol: SERPINB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005697305.3 | XP_005697362.1 | 6 - 7 | 874 | 70 | |
GenBank mRNA | KA339712.1 | - | 6 - 7 | 858 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
