Assay Details
| Gene Symbol: BAHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054445.1 | XP_017909934.1 | 3 - 4 | 1990 | 63 | |
| XM_018054446.1 | XP_017909935.1 | 3 - 4 | 1987 | 63 | ||
| XM_018054447.1 | XP_017909936.1 | 3 - 4 | 1990 | 63 | ||
| GenBank mRNA | JO558412.2 | - | 1 - 2 | 45 | 63 | |
| JT679396.1 | - | 4 - 5 | 1825 | 63 | ||
More Information
Set Membership: |
|

BAHD1
Genomic Map