Assay Details
| Gene Symbol: IL18R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013967547.2 | XP_013823001.2 | 2 - 3 | 656 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IL18R1| Gene Symbol: IL18R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013967547.2 | XP_013823001.2 | 2 - 3 | 656 | 72 | |
Set Membership: |
|