Assay Details
| Gene Symbol: CA14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677683.3 | XP_005677740.1 | 9 - 10 | 1697 | 60 | |
| XM_013962447.2 | XP_013817901.1 | 9 - 10 | 1691 | 60 | ||
| XM_013962448.2 | XP_013817902.1 | 10 - 11 | 1374 | 60 | ||
More Information
Set Membership: |
|

CA14
Genomic Map