Assay Details
Gene Symbol: LOC102184693 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005679940.3 | XP_005679997.1 | 7 - 8 | 1364 | 70 | |
GenBank mRNA | KA330058.1 | - | 2 - 3 | 1538 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
