Assay Details
Gene Symbol: LOC102184693 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005679940.3 | XP_005679997.1 | 8 - 9 | 1401 | 115 | |
GenBank mRNA | KA330058.1 | - | 1 - 2 | 1501 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
