Assay Details
Gene Symbol: CD69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680868.2 | XP_005680925.1 | 3 - 4 | 465 | 76 | |
GenBank mRNA | JO605363.2 | - | 3 - 4 | 519 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
