Assay Details
Gene Symbol: RAD21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058370.1 | XP_017913859.1 | 11 - 12 | 1746 | 62 | |
XM_018058371.1 | XP_017913860.1 | 11 - 12 | 1708 | 62 | ||
GenBank mRNA | JO393556.2 | - | 5 - 6 | 719 | 62 | |
JO580138.1 | - | 11 - 12 | 1689 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
