Assay Details
| Gene Symbol: TPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689574.3 | XP_005689631.2 | 10 - 11 | 1211 | 66 | |
| XM_005689575.3 | XP_005689632.2 | 9 - 10 | 1143 | 66 | ||
| GenBank mRNA | JO413328.2 | - | 3 - 4 | 2074 | 66 | |
| JO413329.2 | - | 3 - 4 | 2074 | 66 | ||
More Information
Set Membership: |
|

TPH1
Genomic Map