Assay Details
Gene Symbol: EGLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062448.1 | XP_017917937.1 | 5 - 6 | 1496 | 59 | |
XM_018062449.1 | XP_017917938.1 | 5 - 6 | 1592 | 59 | ||
GenBank mRNA | JT746028.1 | - | 7 - 8 | 1445 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
