Assay Details
| Gene Symbol: C19H17orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694032.2 | XP_005694089.1 | 2 - 3 | 593 | 58 | |
| XM_018065402.1 | XP_017920891.1 | 2 - 3 | 580 | 58 | ||
| XM_018065403.1 | XP_017920892.1 | 2 - 3 | 580 | 58 | ||
More Information
Set Membership: |
|

C19H17orf62
Genomic Map