Assay Details
Gene Symbol: NAA50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038952.1 | XP_017894441.1 | 2 - 3 | 163 | 62 | |
XM_018039011.1 | XP_017894500.1 | 2 - 3 | 160 | 62 | ||
XM_018039062.1 | XP_017894551.1 | 2 - 3 | 483 | 62 | ||
XM_018039131.1 | XP_017894620.1 | 2 - 3 | 481 | 62 | ||
GenBank mRNA | JO334799.2 | - | 3 - 4 | 445 | 62 | |
JO334800.2 | - | 3 - 4 | 445 | 62 | ||
JO347969.2 | - | 3 - 4 | 1854 | 62 | ||
JO376905.2 | - | 502 | 62 | |||
JO412776.2 | - | 3 - 4 | 512 | 62 | ||
JO412777.2 | - | 3 - 4 | 512 | 62 | ||
JO450251.2 | - | 3 - 4 | 503 | 62 | ||
JO450358.2 | - | 3 - 4 | 503 | 62 | ||
JO508761.2 | - | 3 - 4 | 504 | 62 | ||
JO537751.2 | - | 3 - 4 | 2543 | 62 | ||
JO537752.2 | - | 3 - 4 | 2543 | 62 | ||
JO537753.2 | - | 3 - 4 | 2543 | 62 | ||
JO612888.2 | - | 3 - 4 | 515 | 62 | ||
JO623739.2 | - | 3 - 4 | 533 | 62 | ||
KA340250.1 | - | 2 - 3 | 550 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
