Assay Details
| Gene Symbol: CD74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050158.1 | XP_017905647.1 | 2 - 3 | 459 | 57 | |
| XM_018050159.1 | XP_017905648.1 | 2 - 3 | 459 | 57 | ||
| GenBank mRNA | JO581664.1 | - | 2746 | 57 | ||
More Information
Set Membership: |
|

CD74
Genomic Map